2-Fluoro-1-(methylsulfanyl)-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

34357-5G 34357-25G 34357-1G 34357-500MG
77632-860EA 983 USD
77632-860 77655-234 77646-230 77646-232
2-Fluoro-1-(methylsulfanyl)-3-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-(methylsulphanyl)-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 210.192 g/mol
MW: C₈H₆F₄S
Storage Temperature: Ambient
MDL Number: MFCD22489182
CAS Number: 1824423-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR