3-Bromo-5-chloro-4-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

34286-5G 34286-25G 34286-1G 34286-500MG
77634-868EA 584 USD
77634-868 77658-852 77642-232 77646-226
3-Bromo-5-chloro-4-fluorobenzotrifluoride ≥97%
3-Bromo-5-chloro-4-fluorobenzotrifluoride

Precaución: For Research Use Only

Formula: 277.441 g/mol
MW: C₇H₂BrClF₄
Storage Temperature: Ambient
MDL Number: MFCD32902584
CAS Number: 109919-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR