Pyridine, 4-bromo-2-(2,2-difluoroethoxy)- ≥95%
Supplier: AOBChem USA
|
Warning
|
341160-5G
341160-25G
341160-1G
341160-500MG
77628-716EA
1738.64
USD
77628-716
77635-674
77642-228
77646-218
Pyridine, 4-bromo-2-(2,2-difluoroethoxy)- ≥95%
Pyridine, 4-bromo-2-(2,2-difluoroethoxy)-
Precaución: For Research Use Only
Formula:
238.03 g/mol MW: C₇H₆BrF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD35128214 CAS Number: 2930725-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...