1-Fluoro-3-(methoxymethoxy)-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

34114-5G 34114-1G 34114-25G 34114-500MG
77634-862EA 288 USD
77634-862 77638-226 77655-224 77646-216
1-Fluoro-3-(methoxymethoxy)-5-methylbenzene ≥95%
1-Fluoro-3-(methoxymethoxy)-5-methylbenzene

Precaución: For Research Use Only

Formula: 170.181 g/mol
MW: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD31805769
CAS Number: 2271442-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR