Ethyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34112-5G 34112-25G 34112-1G 34112-500MG
77634-860EA 255 USD
77634-860 77658-848 77642-226 77646-214
Ethyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%
Ethyl 3-bromo-4-chloro-5-fluorobenzoate

Precaución: For Research Use Only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD11846090
CAS Number: 1160574-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR