3,5-Dichloro-2-methyl-benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

34099-5G 34099-25G 34099-1G 34099-500MG
77632-856EA 1134 USD
77632-856 77658-846 77642-224 77650-242
3,5-Dichloro-2-methyl-benzoic acid methyl ester ≥97%
3,5-Dichloro-2-methyl-benzoic acid methyl ester

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24323019
CAS Number: 99849-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR