3-Pyridinamine, 2-(4-chlorophenyl)- ≥97%

Supplier: AOBChem USA

Warning

340926-5G 340926-1G 340926-25G 340926-500MG
77634-858EA 482 USD
77634-858 77638-224 77658-844 77650-240
3-Pyridinamine, 2-(4-chlorophenyl)- ≥97%
3-Pyridinamine, 2-(4-chlorophenyl)-

Precaución: For Research Use Only

Formula: C₁₁H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06740311
CAS Number: 886507-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR