4-Bromo-2-isopropyl-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

34083-5G 34083-25G 34083-1G 34083-500MG
77632-854EA 1258 USD
77632-854 77658-842 77642-222 77646-212
4-Bromo-2-isopropyl-1-methylbenzene ≥97%
4-Bromo-2-isopropyl-1-methylbenzene

Precaución: For Research Use Only

Formula: 213.114 g/mol
MW: C₁₀H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD28356488
CAS Number: 4534-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR