3,5-Difluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

34042-5G 34042-1G 34042-25G 34042-500MG
77632-852EA 995 USD
77632-852 77638-220 77658-840 77646-208
3,5-Difluoro-4'-methyl-1,1'-biphenyl ≥97%
3,5-Difluoro-4'-methyl-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 204.215 g/mol
MW: C₁₃H₁₀F₂
Storage Temperature: Ambient
MDL Number: MFCD31619462
CAS Number: 97067-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR