2-Fluoro-5-(4-methylphenyl)-pyridine ≥95%

Supplier: AOBChem USA

Warning

34037-5G 34037-1G 34037-500MG
77628-714EA 1553.24 USD
77628-714 77638-216 77650-236
2-Fluoro-5-(4-methylphenyl)-pyridine ≥95%
2-Fluoro-5-(4-methylphenyl)-pyridine

Precaución: For Research Use Only

Formula: 187.213 g/mol
MW: C₁₂H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD31619458
CAS Number: 1337563-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR