Triethyl(2-fluoro-5-methylphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

340159-5G 340159-25G 340159-1G 340159-500MG
77628-712EA 1484.23 USD
77628-712 77635-672 77638-214 77646-202
Triethyl(2-fluoro-5-methylphenyl)silane ≥95%
Triethyl(2-fluoro-5-methylphenyl)silane

Precaución: For Research Use Only

Formula: 224.3897 g/mol
MW: C₁₃H₂₁FSi
Storage Temperature: Ambient
MDL Number: MFCD35185172
CAS Number: 3002439-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR