(Z)-1,1,1-Trifluoro-4-hydroxy-4-phenylbut-3-en-2-one ≥95%

Supplier: AOBChem USA

Warning

340144-5G 340144-1G 340144-25G 340144-500MG
77628-710EA 1484.23 USD
77628-710 77638-212 77658-836 77650-234
(Z)-1,1,1-Trifluoro-4-hydroxy-4-phenylbut-3-en-2-one ≥95%
(Z)-1,1,1-Trifluoro-4-hydroxy-4-phenylbut-3-en-2-one

Precaución: For Research Use Only

Formula: 216.1565 g/mol
MW: C₁₀H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28390541
CAS Number: 202334-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR