1-(5-Bromo-2-fluoro-3-hydroxyphenyl)ethan-1-one ≥95%

Supplier: AOBChem USA

Warning

33959-1G 33959-500MG 33959-5G
77634-852EA 1158 USD
77634-852 77652-692 77655-216
1-(5-Bromo-2-fluoro-3-hydroxyphenyl)ethan-1-one ≥95%
1-(5-Bromo-2-fluoro-3-hydroxyphenyl)ethan-1-one

Precaución: For Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738816
CAS Number: 1804906-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR