4-Bromo-2-methoxy-6-methylbenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

33949-5G 33949-1G 33949-500MG 33949-25G
77632-846EA 988 USD
77632-846 77638-208 77652-690 77655-214
4-Bromo-2-methoxy-6-methylbenzoic acid methyl ester ≥97%
4-Bromo-2-methoxy-6-methylbenzoic acid methyl ester

Precaución: For Research Use Only

Formula: 259.097 g/mol
MW: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD23707500
CAS Number: 877149-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR