4-Bromo-2-methoxy-6-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

33947-5G 33947-25G 33947-1G 33947-500MG
77632-844EA 1232 USD
77632-844 77655-212 77642-218 77650-230
4-Bromo-2-methoxy-6-methylbenzonitrile ≥95%
4-Bromo-2-methoxy-6-methylbenzonitrile

Precaución: For Research Use Only

Formula: C₉H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD19105196
CAS Number: 877149-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR