4-Iodo-5-methylpyridin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

33935-5G 33935-1G 33935-25G 33935-500MG
77632-842EA 1430 USD
77632-842 77638-206 77655-210 77650-228
4-Iodo-5-methylpyridin-2(1H)-one ≥97%
4-Iodo-5-methylpyridin-2(1H)-one

Precaución: For Research Use Only

Formula: 235.022 g/mol
MW: C₆H₆INO
Storage Temperature: Ambient
MDL Number: MFCD25956808
CAS Number: 1227570-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR