3,4'-Bis(trifluoromethyl)benzophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(Trifluormethyl)-3-((4-(trifluormethyl)phenyl)methanoyl)benzene

339251-5G 339251-25G 339251-1G 339251-500MG
77634-850EA 343 USD
77634-850 77658-832 77642-216 77650-226
3,4'-Bis(trifluoromethyl)benzophenone ≥97%
3,4'-Bis(trifluoromethyl)benzophenone

Precaución: For Research Use Only

Formula: C₁₅H₈F₆O
MW: 318.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013561
CAS Number: 21084-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR