6-Chloro-N-methoxy-N-methylnicotinamide ≥95%
Supplier: AOBChem USA
|
Warning
|
33923-5G
33923-1G
33923-25G
77634-848EA
200
USD
77634-848
77638-204
77655-208
6-Chloro-N-methoxy-N-methylnicotinamide ≥95%
6-Chloro-N-methoxy-N-methylnicotinamide
Precaución: For Research Use Only
Formula:
C₈H₉ClN₂O₂ MW: 200.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09991647 CAS Number: 149281-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...