1-(6-Methoxypyridin-3-yl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

33921-5G 33921-25G 33921-1G 33921-500MG
77632-840EA 1030 USD
77632-840 77655-206 77642-214 77646-200
1-(6-Methoxypyridin-3-yl)propan-1-one ≥97%
1-(6-Methoxypyridin-3-yl)propan-1-one

Precaución: For Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18802862
CAS Number: 885229-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR