4-Propionylbiphenyl ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Phenylpropiophenone

339192-5G 339192-25G
77634-846EA 86 USD
77634-846 77655-204
4-Propionylbiphenyl ≥97%
4-Propionylbiphenyl

Precaución: For Research Use Only

Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027015
CAS Number: 37940-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR