1-(4-(Trifluoromethoxy)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

338939-5G 338939-1G 338939-500MG
77628-706EA 1484.23 USD
77628-706 77642-212 77646-198
1-(4-(Trifluoromethoxy)phenyl)prop-2-en-1-ol ≥95%
1-(4-(Trifluoromethoxy)phenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 218.172 g/mol
MW: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD21232139
CAS Number: 1476730-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR