2-(4-(Benzyloxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

33875-5G 33875-25G 33875-1G 33875-500MG
77634-844EA 483 USD
77634-844 77658-830 77642-206 77646-194
2-(4-(Benzyloxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For Research Use Only

Formula: C₂₃H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD28505152
CAS Number: 2096997-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR