Cyclopropyl(2,3,5,6-tetrafluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

338603-5G 338603-1G 338603-500MG
77628-698EA 1484.23 USD
77628-698 77638-202 77646-190
Cyclopropyl(2,3,5,6-tetrafluorophenyl)methanol ≥95%
Cyclopropyl(2,3,5,6-tetrafluorophenyl)methanol

Precaución: For Research Use Only

Formula: 220.164 g/mol
MW: C₁₀H₈F₄O
Storage Temperature: Ambient
CAS Number: 1858045-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR