3-(3,5-Di-tert-butylphenyl)oxetan-3-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
338588-5G
338588-1G
338588-25G
338588-500MG
77628-696EA
1484.23
USD
77628-696
77638-200
77655-198
77646-188
3-(3,5-Di-tert-butylphenyl)oxetan-3-ol ≥95%
3-(3,5-Di-tert-butylphenyl)oxetan-3-ol
Precaución: For Research Use Only
Formula:
C₁₇H₂₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD35184219 CAS Number: 3002479-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...