1-(3,5-Di-tert-butylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

338573-5G 338573-25G 338573-1G 338573-500MG
77628-694EA 1484.23 USD
77628-694 77655-196 77646-184 77646-186
1-(3,5-Di-tert-butylphenyl)cyclopentanol ≥95%
1-(3,5-Di-tert-butylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₉H₃₀O
Storage Temperature: Ambient
MDL Number: MFCD35184205
CAS Number: 3002504-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR