(3,5-Di-tert-butylphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

338566-5G 338566-25G 338566-1G 338566-500MG
77632-838EA 1441 USD
77632-838 77658-828 77642-204 77646-182
(3,5-Di-tert-butylphenyl)(phenyl)methanone ≥95%
(3,5-Di-tert-butylphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₂₁H₂₆O
Storage Temperature: Ambient
MDL Number: MFCD35184198
CAS Number: 3002429-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR