(4-Methoxy-3-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
338479-5G
338479-25G
338479-1G
338479-500MG
77632-836EA
1441
USD
77632-836
77655-194
77642-202
77650-220
(4-Methoxy-3-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%
(4-Methoxy-3-(trifluoromethyl)phenyl)(phenyl)methanone
Precaución: For Research Use Only
Formula:
280.242 g/mol MW: C₁₅H₁₁F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD35184122 CAS Number: 2169685-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...