1-(4-Methoxy-3-(trifluoromethyl)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

338477-5G 338477-1G 338477-25G 338477-500MG
77628-692EA 1484.23 USD
77628-692 77638-198 77655-192 77646-180
1-(4-Methoxy-3-(trifluoromethyl)phenyl)cyclopentanol ≥95%
1-(4-Methoxy-3-(trifluoromethyl)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 260.252 g/mol
MW: C₁₃H₁₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35184120
CAS Number: 3002443-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR