2-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

338436-5G 338436-25G 338436-1G 338436-500MG
77628-688EA 1484.23 USD
77628-688 77655-190 77642-198 77646-178
2-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 292.36 g/mol
MW: C₁₄H₁₉F₃OS
Storage Temperature: Ambient
MDL Number: MFCD35184085
CAS Number: 3002506-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR