1-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

338427-5G 338427-25G 338427-1G 338427-500MG
77628-686EA 1484.23 USD
77628-686 77655-188 77642-196 77650-216
1-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)ethanone ≥95%
1-(4-(Tert-butylthio)-3-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 276.318 g/mol
MW: C₁₃H₁₅F₃OS
Storage Temperature: Ambient
MDL Number: MFCD35184076
CAS Number: 3002500-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR