Tert-butyl(4-(methylthio)-2-(trifluoromethyl)phenyl)sulphane ≥95%

Supplier: AOBChem USA

Warning

338426-5G 338426-25G 338426-1G 338426-500MG
77628-684EA 1501.74 USD
77628-684 77635-668 77642-194 77646-176
Tert-butyl(4-(methylthio)-2-(trifluoromethyl)phenyl)sulphane ≥95%
Tert-butyl(4-(methylthio)-2-(trifluoromethyl)phenyl)sulphane

Precaución: For Research Use Only

Formula: C₁₂H₁₅F₃S₂
Storage Temperature: Ambient
MDL Number: MFCD35184075
CAS Number: 3002443-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR