4-(Tert-butylthio)-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

338424-5G 338424-25G 338424-1G 338424-500MG
77628-682EA 1777.78 USD
77628-682 77635-666 77638-194 77646-172
4-(Tert-butylthio)-3-(trifluoromethyl)benzoic acid ≥97%
4-(Tert-butylthio)-3-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: 278.291 g/mol
MW: C₁₂H₁₃F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD35184073
CAS Number: 3002482-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR