2,6-Dichloro-4-(methylsulfanyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

33834-5G 33834-1G 33834-25G 33834-500MG
77634-842EA 400 USD
77634-842 77638-192 77655-186 77650-214
2,6-Dichloro-4-(methylsulfanyl)benzoic acid ≥95%
2,6-Dichloro-4-(methylsulphanyl)benzoic acid

Precaución: For Research Use Only

Formula: 237.103 g/mol
MW: C₈H₆Cl₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD31561339
CAS Number: 2145093-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR