Methyl 3-cyano-2,4-dichlorobenzoate ≥95%

Supplier: AOBChem USA

Warning

33772-5G 33772-1G 33772-500MG
77628-676EA 1612.98 USD
77628-676 77642-190 77646-170
Methyl 3-cyano-2,4-dichlorobenzoate ≥95%
Methyl 3-cyano-2,4-dichlorobenzoate

Precaución: For Research Use Only

Formula: C₉H₅Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD28758644
CAS Number: 198273-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR