1,3-Bis(benzyloxy)-5-bromobenzene ≥95%

Supplier: AOBChem USA

Warning

33764-5G 33764-1G 33764-25G 33764-500MG
77634-840EA 399 USD
77634-840 77638-186 77655-182 77646-168
1,3-Bis(benzyloxy)-5-bromobenzene ≥95%
1,3-Bis(benzyloxy)-5-bromobenzene

Precaución: For Research Use Only

Formula: 369.252 g/mol
MW: C₂₀H₁₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD26131152
CAS Number: 128924-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR