4-Chloro-3-fluoro-2-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33730-5G 33730-25G 33730-1G 33730-500MG
77632-830EA 1258 USD
77632-830 77655-180 77642-184 77650-206
4-Chloro-3-fluoro-2-hydroxybenzoic acid ≥95%
4-Chloro-3-fluoro-2-hydroxybenzoic acid

Precaución: For Research Use Only

Formula: C₇H₄ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD22415148
CAS Number: 1702676-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR