1-Bromo-5-(4-fluorophenyl)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

33712-5G 33712-25G 33712-1G 33712-500MG
77632-828EA 871 USD
77632-828 77655-178 77642-182 77650-204
1-Bromo-5-(4-fluorophenyl)-2-methylbenzene ≥95%
1-Bromo-5-(4-fluorophenyl)-2-methylbenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD31561317
CAS Number: 2145093-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR