2-Bromo-4,5-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33695-5G 33695-1G 33695-25G 33695-500MG
77634-838EA 468 USD
77634-838 77638-182 77658-820 77650-202
2-Bromo-4,5-dimethylbenzoic acid ≥95%
2-Bromo-4,5-dimethylbenzoic acid

Precaución: For Research Use Only

Formula: 229.071 g/mol
MW: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16658606
CAS Number: 4612-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR