1-Bromo-3-chloro-2-((4-methylbenzyl)oxy)benzene ≥95%

Supplier: AOBChem USA

Warning

336861-5G 336861-25G 336861-1G 336861-500MG
77628-668EA 1484.23 USD
77628-668 77658-818 77642-180 77650-200
1-Bromo-3-chloro-2-((4-methylbenzyl)oxy)benzene ≥95%
1-Bromo-3-chloro-2-((4-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 311.601 g/mol
MW: C₁₄H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD35183304
CAS Number: 3002485-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR