3-Bromo-2,5-dichlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33672-5G 33672-25G 33672-1G 33672-500MG
77634-836EA 468 USD
77634-836 77655-176 77642-178 77646-164
3-Bromo-2,5-dichlorophenylboronic acid pinacol ester ≥97%
3-Bromo-2,5-dichlorophenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 351.859 g/mol
MW: C₁₂H₁₄BBrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31382568
CAS Number: 2121515-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR