N-(2,6-Diethylphenyl)maleimide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,6-Diethylphenyl)-1H-pyrrole-2,5-dione

336584-5G 336584-25G 336584-1G
77634-834EA 180 USD
77634-834 77655-174 77642-176
N-(2,6-Diethylphenyl)maleimide ≥97%
N-(2,6-Diethylphenyl)maleimide

Precaución: For Research Use Only

Formula: C₁₄H₁₅NO₂
MW: 229.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173826
CAS Number: 38167-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR