Methyl 3-bromo-2,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

33655-5G 33655-1G 33655-25G 33655-500MG
77632-826EA 793 USD
77632-826 77638-180 77658-816 77650-198
Methyl 3-bromo-2,5-dichlorobenzoate ≥97%
Methyl 3-bromo-2,5-dichlorobenzoate

Precaución: For Research Use Only

Formula: C₈H₅BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07780741
CAS Number: 933585-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR