2,4-Dimethyl-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33643-5G 33643-1G 33643-25G 33643-500MG
77632-822EA 942 USD
77632-822 77638-178 77655-168 77650-196
2,4-Dimethyl-3-fluorobenzoic acid ≥97%
2,4-Dimethyl-3-fluorobenzoic acid

Precaución: For Research Use Only

Formula: 168.165 g/mol
MW: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD00184981
CAS Number: 26583-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR