3-Fluoro-2,4-dimethylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

33640-5G 33640-25G 33640-1G 33640-500MG
77632-820EA 1323 USD
77632-820 77655-166 77642-170 77646-160
3-Fluoro-2,4-dimethylphenylboronic acid pinacol ester ≥95%
3-Fluoro-2,4-dimethylphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 250.117 g/mol
MW: C₁₄H₂₀BFO₂
Storage Temperature: Ambient
MDL Number: MFCD31561307
CAS Number: 2819649-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR