4-Bromo-2,5-difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

33625-5G 33625-25G 33625-1G 33625-500MG
77634-830EA 425 USD
77634-830 77655-164 77642-168 77650-194
4-Bromo-2,5-difluoro-1,1'-biphenyl ≥97%
4-Bromo-2,5-difluoro-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 269.085 g/mol
MW: C₁₂H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD31561306
CAS Number: 121219-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR