3-(Benzyloxy)-6-bromo-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

33586-5G 33586-1G 33586-25G 33586-500MG
77632-816EA 664 USD
77632-816 77638-174 77658-814 77650-190
3-(Benzyloxy)-6-bromo-2-methylpyridine ≥97%
3-(Benzyloxy)-6-bromo-2-methylpyridine

Precaución: For Research Use Only

Formula: 278.144 g/mol
MW: C₁₃H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD31382555
CAS Number: 2121515-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR