(2,5-Dichloro-3-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

33571-5G 33571-1G 33571-25G 33571-500MG
77632-814EA 1289 USD
77632-814 77638-172 77658-810 77646-154
(2,5-Dichloro-3-fluorophenyl)methanol ≥97%
(2,5-Dichloro-3-fluorophenyl)methanol

Precaución: For Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28524129
CAS Number: 1615212-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR