2,5-Dichloro-3-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33568-5G 33568-25G 33568-1G 33568-500MG
77632-812EA 841 USD
77632-812 77655-156 77642-160 77650-186
2,5-Dichloro-3-fluorophenylboronic acid pinacol ester ≥97%
2,5-Dichloro-3-fluorophenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 290.954 g/mol
MW: C₁₂H₁₄BCl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD22414473
CAS Number: 2121515-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR