Methyl 2-chloro-5-(trifluoromethoxy)benzoate ≥95%

Supplier: AOBChem USA

Warning

33551-5G 33551-25G 33551-1G 33551-500MG
77634-822EA 536 USD
77634-822 77658-808 77642-158 77646-152
Methyl 2-chloro-5-(trifluoromethoxy)benzoate ≥95%
Methyl 2-chloro-5-(trifluoromethoxy)benzoate

Precaución: For Research Use Only

Formula: C₉H₆ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18399022
CAS Number: 1261649-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR