1-Bromo-2-fluoro-3-methoxy-5-(trifluoromethyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
33512-5G
33512-25G
33512-1G
33512-500MG
77634-818EA
590
USD
77634-818
77658-806
77642-152
77650-182
1-Bromo-2-fluoro-3-methoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-2-fluoro-3-methoxy-5-(trifluoromethyl)benzene
Precaución: For Research Use Only
Formula:
273.022 g/mol MW: C₈H₅BrF₄O Storage Temperature: Ambient |
MDL Number:
MFCD32631804 CAS Number: 2091605-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...